Assay Details
Gene Symbol: TRAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666444.3 | XP_005666501.1 | 2515 | 85 | ||
GenBank mRNA | AK346671.1 | - | 211 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005666444.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
