Assay Details
Gene Symbol: BCL7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124483.5 | XP_003124531.1 | 622 | 90 | ||
XM_013995586.2 | XP_013851040.1 | 633 | 90 | |||
GenBank mRNA | AK346920.1 | - | 4 - 5 | 597 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
