Assay Details
Gene Symbol: RPS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125136.5 | XP_003125184.3 | 195 | 124 | ||
GenBank mRNA | AK346930.1 | - | 3 - 4 | 137 | 124 | |
AK391678.1 | - | 3 - 4 | 144 | 124 | ||
AK391997.1 | - | 3 - 4 | 138 | 124 | ||
AK399952.1 | - | 3 - 4 | 157 | 124 | ||
AY610307.1 | - | 3 - 4 | 134 | 124 | ||
AY610373.1 | - | 2 - 3 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
