Assay Details
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087480.1 | XP_020943139.1 | 474 | 139 | ||
GenBank mRNA | AK347013.1 | - | 2 - 3 | 464 | 139 | |
AK391841.1 | - | 2 - 3 | 462 | 139 | ||
AK400030.1 | - | 2 - 3 | 481 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
