Assay Details
Gene Symbol: UBL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003360512.4 | XP_003360560.1 | 574 | 85 | ||
GenBank mRNA | AK347248.1 | - | 4 - 5 | 527 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
