Assay Details
Gene Symbol: IL17RC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069809.1 | XP_020925468.1 | 433 | 67 | ||
GenBank mRNA | AK347630.1 | - | 6 | 849 | 67 | |
AK396665.1 | - | 6 | 844 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
