Assay Details
Gene Symbol: FAM234A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003481091.4 | XP_003481139.1 | 1481 | 66 | ||
XM_005655119.3 | XP_005655176.1 | 1389 | 66 | |||
XM_005655121.3 | XP_005655178.1 | 1490 | 66 | |||
XM_005655122.3 | XP_005655179.1 | 1358 | 66 | |||
XM_005655123.3 | XP_005655180.1 | 1290 | 66 | |||
XM_013987739.2 | XP_013843193.1 | 1395 | 66 | |||
XM_013987740.2 | XP_013843194.1 | 1363 | 66 | |||
XM_021086932.1 | XP_020942591.1 | 1571 | 66 | |||
XM_021086933.1 | XP_020942592.1 | 1692 | 66 | |||
XM_021086934.1 | XP_020942593.1 | 1363 | 66 | |||
XM_021086935.1 | XP_020942594.1 | 1394 | 66 | |||
XM_021086936.1 | XP_020942595.1 | 1363 | 66 | |||
GenBank mRNA | AK347646.1 | - | 10 - 11 | 1453 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
