Assay Details
Gene Symbol: TBCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021084475.1 | XP_020940134.1 | 342 | 82 | ||
GenBank mRNA | AK347697.1 | - | 3 - 4 | 324 | 82 | |
AK392112.1 | - | 3 - 4 | 307 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
