Assay Details
Gene Symbol: TRIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013996340.2 | XP_013851794.2 | 663 | 122 | ||
GenBank mRNA | AK348679.1 | - | 1 | 524 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013996340.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
