Assay Details
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021067910.1 | XP_020923569.1 | 1960 | 61 | ||
GenBank mRNA | AK349146.1 | - | 3 - 4 | 678 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
