Assay Details
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021067910.1 | XP_020923569.1 | 2189 | 72 | ||
GenBank mRNA | AK349146.1 | - | 4 - 5 | 907 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
