Assay Details
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131420.5 | XP_003131468.2 | 989 | 95 | ||
XM_021066991.1 | XP_020922650.1 | 960 | 95 | |||
XM_021066992.1 | XP_020922651.1 | 1048 | 95 | |||
GenBank mRNA | AK349242.1 | - | 3 - 4 | 929 | 95 | |
AK390110.1 | - | 3 - 4 | 903 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
