Assay Details
Gene Symbol: CYBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021066352.1 | XP_020922011.1 | 432 | 103 | ||
GenBank mRNA | AK349285.1 | - | 3 - 4 | 401 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021066352.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
