Assay Details
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001926447.5 | XP_001926482.2 | 1230 | 66 | ||
XM_005670996.3 | XP_005671053.1 | 1163 | 66 | |||
XM_005670997.3 | XP_005671054.1 | 1643 | 66 | |||
XM_021072775.1 | XP_020928434.1 | 1476 | 66 | |||
XM_021072776.1 | XP_020928435.1 | 1648 | 66 | |||
XM_021072777.1 | XP_020928436.1 | 1459 | 66 | |||
XM_021072778.1 | XP_020928437.1 | 1402 | 66 | |||
XM_021072779.1 | XP_020928438.1 | 1253 | 66 | |||
XM_021072780.1 | XP_020928439.1 | 3647 | 66 | |||
GenBank mRNA | AK349315.1 | - | 10 - 11 | 1198 | 66 | |
AK390297.1 | - | 9 - 10 | 1245 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
