Assay Details
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021084255.1 | XP_020939914.1 | 1641 | 66 | ||
XM_021084257.1 | XP_020939916.1 | 1638 | 66 | |||
XM_021084258.1 | XP_020939917.1 | 1641 | 66 | |||
XM_021084259.1 | XP_020939918.1 | 1637 | 66 | |||
GenBank mRNA | AK349349.1 | - | 7 - 8 | 829 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
