Assay Details
Gene Symbol: STMN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021077837.1 | XP_020933496.1 | 1103 | 61 | ||
GenBank mRNA | AK349408.1 | - | 1 - 2 | 155 | 61 | |
AK392217.1 | - | 1 - 2 | 88 | 61 | ||
AK399985.1 | - | 1 - 2 | 172 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021077837.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
