Assay Details
Gene Symbol: STMN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021077837.1 | XP_020933496.1 | 1569 | 106 | ||
GenBank mRNA | AK349408.1 | - | 4 - 5 | 621 | 106 | |
AK392217.1 | - | 4 - 5 | 554 | 106 | ||
AK399985.1 | - | 4 - 5 | 638 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
