Assay Details
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126415.4 | XP_003126463.1 | 362 | 93 | ||
GenBank mRNA | AK349478.1 | - | 1 - 2 | 336 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
