Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001925572.5 | XP_001925607.3 | 710 | 72 | ||
GenBank mRNA | AK349512.1 | - | 2 - 3 | 605 | 72 | |
AK400347.1 | - | 2 - 3 | 688 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
