Assay Details
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126802.4 | XP_003126850.1 | 362 | 71 | ||
GenBank mRNA | AK349577.1 | - | 3 - 4 | 811 | 71 | |
AK396881.1 | - | 3 - 4 | 335 | 71 | ||
AK399792.1 | - | 3 - 4 | 303 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
