Assay Details
Gene Symbol: JHY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005667440.2 | XP_005667497.2 | 1094 | 96 | ||
XM_013979491.2 | XP_013834945.2 | 1063 | 96 | |||
GenBank mRNA | AK349582.1 | - | 3 - 4 | 1094 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
