Assay Details
Gene Symbol: PRSS27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086748.1 | XP_020942407.1 | 400 | 109 | ||
XM_021086750.1 | XP_020942409.1 | 408 | 109 | |||
GenBank mRNA | AK349701.1 | - | 2 - 3 | 148 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
