Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013978589.2 | XP_013834043.2 | 605 | 62 | ||
GenBank mRNA | AK350273.1 | - | 5 - 6 | 699 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
