Assay Details
Gene Symbol: CDKN1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021082622.1 | XP_020938281.1 | 381 | 69 | ||
GenBank mRNA | AK350718.1 | - | 1 - 2 | 153 | 69 | |
AK395505.1 | - | 1 - 2 | 142 | 69 | ||
HQ679903.1 | - | 1 - 2 | 151 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021082622.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
