Assay Details
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003360516.3 | XP_003360564.1 | 648 | 61 | ||
XM_005674056.3 | XP_005674113.1 | 648 | 61 | |||
XM_021080750.1 | XP_020936409.1 | 648 | 61 | |||
XM_021080751.1 | XP_020936410.1 | 718 | 61 | |||
XM_021080752.1 | XP_020936411.1 | 758 | 61 | |||
GenBank mRNA | AK351158.1 | - | 5 - 6 | 623 | 61 | |
AK351160.1 | - | 5 - 6 | 630 | 61 | ||
AK351266.1 | - | 5 - 6 | 618 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
