Assay Details
Gene Symbol: FYB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096577.1 | XP_020952236.1 | 1779 | 108 | ||
XM_021096578.1 | XP_020952237.1 | 1776 | 108 | |||
XM_021096579.1 | XP_020952238.1 | 1779 | 108 | |||
XM_021096580.1 | XP_020952239.1 | 1779 | 108 | |||
GenBank mRNA | AK352268.1 | - | 11 - 12 | 1660 | 108 | |
AK396675.1 | - | 11 - 12 | 1708 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
