Assay Details
Gene Symbol: FYB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096577.1 | XP_020952236.1 | 258 | 101 | ||
XM_021096578.1 | XP_020952237.1 | 258 | 101 | |||
XM_021096579.1 | XP_020952238.1 | 258 | 101 | |||
XM_021096580.1 | XP_020952239.1 | 258 | 101 | |||
GenBank mRNA | AK352268.1 | - | 1 - 2 | 139 | 101 | |
AK396675.1 | - | 1 - 2 | 187 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
