Assay Details
Gene Symbol: FYB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096577.1 | XP_020952236.1 | 1373 | 91 | ||
XM_021096578.1 | XP_020952237.1 | 1370 | 91 | |||
XM_021096579.1 | XP_020952238.1 | 1373 | 91 | |||
XM_021096580.1 | XP_020952239.1 | 1373 | 91 | |||
GenBank mRNA | AK352268.1 | - | 7 - 8 | 1254 | 91 | |
AK396675.1 | - | 7 - 8 | 1302 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
