Assay Details
Gene Symbol: MOB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003130696.6 | XP_003130744.3 | 1455 | 140 | ||
XM_013980202.2 | XP_013835656.2 | 2378 | 140 | |||
GenBank mRNA | AK352323.1 | - | 3 - 4 | 1058 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
