Assay Details
Gene Symbol: MAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124821.6 | XP_003124869.4 | 977 | 168 | ||
GenBank mRNA | AK352334.1 | - | 3 - 4 | 494 | 168 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
