Assay Details
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003357394.4 | XP_003357442.1 | 991 | 80 | ||
GenBank mRNA | AK352446.1 | - | 3 - 4 | 747 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
