Assay Details
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003357394.4 | XP_003357442.1 | 1268 | 86 | ||
GenBank mRNA | AK352446.1 | - | 5 - 6 | 1024 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
