Assay Details
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021083007.1 | XP_020938666.1 | 1814 | 117 | ||
XM_021083008.1 | XP_020938667.1 | 1130 | 117 | |||
GenBank mRNA | AK389239.1 | - | 1 - 2 | 36 | 117 | |
AK395415.1 | - | 9 - 10 | 862 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
