Assay Details
Gene Symbol: ADAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013988486.2 | XP_013843940.1 | 73 | |||
XM_021093734.1 | XP_020949393.1 | 73 | ||||
GenBank mRNA | AK389313.1 | - | 1 - 2 | 91 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
