Assay Details
Gene Symbol: ADAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013988486.2 | XP_013843940.1 | 1731 | 99 | ||
XM_021093734.1 | XP_020949393.1 | 1590 | 99 | |||
GenBank mRNA | AK389313.1 | - | 4 - 5 | 959 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
