Assay Details
Gene Symbol: MRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131810.4 | XP_003131858.1 | 592 | 101 | ||
GenBank mRNA | AK389406.1 | - | 1 - 2 | 198 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
