Assay Details
Gene Symbol: NUDT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003481783.4 | XP_003481831.1 | 282 | 96 | ||
GenBank mRNA | AK231639.1 | - | 2 - 3 | 215 | 96 | |
AK239889.1 | - | 2 - 3 | 180 | 96 | ||
AK397848.1 | - | 2 - 3 | 245 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
