Assay Details
Gene Symbol: TRMT112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122572.5 | XP_003122620.3 | 570 | 105 | ||
GenBank mRNA | AK231682.1 | - | 1 - 2 | 466 | 105 | |
AK233779.1 | - | 1 - 2 | 479 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
