Assay Details
Gene Symbol: CDC42EP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122538.5 | XP_003122586.1 | 574 | 92 | ||
GenBank mRNA | AK231696.1 | - | 1 - 2 | 97 | 92 | |
AK351381.1 | - | 1 - 2 | 77 | 92 | ||
AK395903.1 | - | 1 - 2 | 105 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003122538.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
