Assay Details
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK231759.1 | - | 2 - 3 | 427 | 57 | |
AK347443.1 | - | 2 - 3 | 394 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK231759.1 | - | 2 - 3 | 427 | 57 | |
AK347443.1 | - | 2 - 3 | 394 | 57 |
Set Membership: |
![]() ![]() ![]() ![]() |