Assay Details
Gene Symbol: DYNLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131635.5 | XP_003131683.1 | 74 | |||
GenBank mRNA | AK389935.1 | - | 1 - 2 | 267 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003131635.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
