Assay Details
Gene Symbol: CAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021100214.1 | XP_020955873.1 | 1006 | 86 | ||
GenBank mRNA | AK390360.1 | - | 5 - 6 | 506 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
