Assay Details
Gene Symbol: STK35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005672793.3 | XP_005672850.1 | 2011 | 72 | ||
GenBank mRNA | AK390429.1 | - | 2 - 3 | 733 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
