Assay Details
Gene Symbol: MCF2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069892.1 | XP_020925551.1 | 1696 | 66 | ||
XM_021069894.1 | XP_020925553.1 | 1544 | 66 | |||
XM_021069895.1 | XP_020925554.1 | 1544 | 66 | |||
XM_021069896.1 | XP_020925555.1 | 1723 | 66 | |||
XM_021069897.1 | XP_020925556.1 | 1544 | 66 | |||
XM_021069898.1 | XP_020925557.1 | 1830 | 66 | |||
XM_021069899.1 | XP_020925558.1 | 1060 | 66 | |||
GenBank mRNA | AK390445.1 | - | 11 - 12 | 1724 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
