Assay Details
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021070394.1 | XP_020926053.1 | 790 | 107 | ||
XM_021070395.1 | XP_020926054.1 | 795 | 107 | |||
GenBank mRNA | AK391007.1 | - | 1 - 2 | 281 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
