Assay Details
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021070395.1 | XP_020926054.1 | 1645 | 76 | ||
GenBank mRNA | AK391007.1 | - | 7 - 8 | 1131 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
