Assay Details
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003135180.4 | XP_003135228.1 | 155 | 100 | ||
GenBank mRNA | AK391214.1 | - | 1 - 2 | 98 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135180.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
