Assay Details
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003135180.4 | XP_003135228.1 | 215 | 115 | ||
GenBank mRNA | AK391214.1 | - | 2 - 3 | 158 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
