Assay Details
Gene Symbol: CEBPZOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087632.1 | XP_020943291.1 | 206 | 189 | ||
GenBank mRNA | AK391287.1 | - | 1 - 2 | 145 | 189 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
