Assay Details
Gene Symbol: MRPL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124936.4 | XP_003124984.1 | 246 | 66 | ||
GenBank mRNA | AK391891.1 | - | 1 - 2 | 60 | 66 | |
AY610044.1 | - | 2 - 3 | 195 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
